MAT236031855 Matrix Scientific

Fmoc-HomoArg(Pbf)-OH

  • MDL number MFCD03093571
  • Cas Number : 1159680-21-3
  • Molecular Weight : 662.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/041281.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/041281.html
Availability details for
Quantity
  • Availability
  • Add to Cart