MAT236032076 Matrix Scientific

Fmoc-2-amino-5-iodobenzoic acid

  • MDL number MFCD02682211
  • Cas Number : 924817-95-8
  • Molecular Weight : 485.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041715.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041715.html
Availability details for
Quantity
  • Availability
  • Add to Cart