MAT236032108 Matrix Scientific

Boc-3-exo-aminobicyclo[2.2.1]-heptane-2-exo-carboxylic acid

  • MDL number MFCD02682613
  • Cas Number : 76198-37-3
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/041355.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041355.html
Availability details for
Quantity
  • Availability
  • Add to Cart