MAT236032256 Matrix Scientific

Boc-7-hydroxy-(R)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD00792389
  • Cas Number : 214630-00-9
  • Molecular Weight : 293.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/041895.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041895.html
Availability details for
Quantity
  • Availability
  • Add to Cart