MAT236032317 Matrix Scientific

Fmoc-3-iodo-L-tyrosine

  • MDL number MFCD00235906
  • Cas Number : 134486-00-3
  • Molecular Weight : 529.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/041956.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/041956.html
Availability details for
Quantity
  • Availability
  • Add to Cart