MAT236032393 Matrix Scientific

Boc-cis-4-cyano-L-proline methyl ester

  • MDL number MFCD03787928
  • Cas Number : 487048-28-2
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042032.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042032.html
Availability details for
Quantity
  • Availability
  • Add to Cart