MAT236032424 Matrix Scientific

Fmoc-trans-4-Hydroxy-D-proline

  • MDL number MFCD01862677
  • Cas Number : 139262-20-7
  • Molecular Weight : 353.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/042063.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/042063.html
Availability details for
Quantity
  • Availability
  • Add to Cart