MAT236032461 Matrix Scientific

Fmoc-L-Ser(OBzl)-OH

  • MDL number MFCD00065674
  • Cas Number : 83792-48-7
  • Molecular Weight : 417.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042100.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042100.html
Availability details for
Quantity
  • Availability
  • Add to Cart