MAT236032569 Matrix Scientific

Boc-(R)-3-amino-4-(3,4-dichlorophenyl)butyric acid

  • MDL number MFCD01860949
  • Cas Number : 269396-56-7
  • Molecular Weight : 348.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042208.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042208.html
Availability details for
Quantity
  • Availability
  • Add to Cart