MAT236032670 Matrix Scientific

Boc-O-benzyl-L-beta-homohydroxyproline

  • MDL number MFCD12547722
  • Cas Number : 136271-81-3
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042292.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042292.html
Availability details for
Quantity
  • Availability
  • Add to Cart