MAT236032762 Matrix Scientific

4-(1-Fmoc-piperidine-4-yl)-butanoic acid

  • MDL number MFCD04114964
  • Cas Number : 885274-47-5
  • Molecular Weight : 393.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/042384.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042384.html
Availability details for
Quantity
  • Availability
  • Add to Cart