MAT238558622 Matrix Scientific

4-Hydroxy-2-(trifluoromethyl)pyrimidine

  • MDL number MFCD00151833
  • Cas Number : 1546-80-1
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-171
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/001424.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001424.html
Availability details for
Quantity
  • Availability
  • Add to Cart