MAT238924851 Matrix Scientific

1-(4-Chlorobenzyl)-1H-1,2,3,4-tetraazol-5-ylamine

  • MDL number MFCD01175876
  • Cas Number : 31694-94-7
  • Molecular Weight : 209.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-203
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042499.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042499.html
Availability details for
Quantity
  • Availability
  • Add to Cart