MAT238924855 Matrix Scientific

3-(Dimethylamino)-2-(2-thienyl)acrylonitrile

  • MDL number MFCD00138239
  • Cas Number : 165404-37-5
  • Molecular Weight : 178.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-64
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042503.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042503.html
Availability details for
Quantity
  • Availability
  • Add to Cart