MAT238924860 Matrix Scientific

7-Chloro-2,4-bis(trifluoromethyl)-[1,8]naphthyridine

  • MDL number MFCD00140156
  • Cas Number : 106582-41-6
  • Molecular Weight : 300.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042508.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042508.html
Availability details for
Quantity
  • Availability
  • Add to Cart