MAT238924862 Matrix Scientific

Ethyl 2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD00139510
  • Cas Number : 81448-48-8
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-82
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042510.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042510.html
Availability details for
Quantity
  • Availability
  • Add to Cart