MAT238924884 Matrix Scientific

1-[(Aminooxy)methyl]-3-chlorobenzene hydrochloride

  • MDL number MFCD01114569
  • Cas Number : 29605-78-5
  • Molecular Weight : 194.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-212
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042532.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042532.html
Availability details for
Quantity
  • Availability
  • Add to Cart