MAT238924894 Matrix Scientific

3-[(3,4-Dichlorobenzyl)sulfanyl]-2-thiophenecarboxylic acid

  • MDL number MFCD00663856
  • Cas Number : 251096-84-1
  • Molecular Weight : 319.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042542.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042542.html
Availability details for
Quantity
  • Availability
  • Add to Cart