MAT238924938 Matrix Scientific

1-(2-Furylcarbonyl)-4-hydroxy-2-pyrrolidinecarboxylic acid

  • MDL number MFCD00202703
  • Cas Number : 251310-45-9
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224-227
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042586.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042586.html
Availability details for
Quantity
  • Availability
  • Add to Cart