MAT238924945 Matrix Scientific

5-{[4-(Trifluoromethyl)phenyl]sulfanyl}-1,2,3-thiadiazole-4-carboxylic acid

  • MDL number MFCD01936015
  • Cas Number : 338982-07-3
  • Molecular Weight : 306.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-166
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042593.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042593.html
Availability details for
Quantity
  • Availability
  • Add to Cart