MAT238924956 Matrix Scientific

2-Chloro-6-(4-fluorophenyl)nicotinonitrile

  • MDL number MFCD00215072
  • Cas Number : 31776-83-7
  • Molecular Weight : 232.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042604.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042604.html
Availability details for
Quantity
  • Availability
  • Add to Cart