MAT238924982 Matrix Scientific

6-Amino-5-chloronicotinonitrile

  • MDL number MFCD03617768
  • Cas Number : 156361-02-3
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-188
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042630.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042630.html
Availability details for
Quantity
  • Availability
  • Add to Cart