MAT238925001 Matrix Scientific

Methyl 4-amino-2-hydroxybenzenecarboxylate

  • MDL number MFCD00088091
  • Cas Number : 4136-97-4
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-123
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042649.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042649.html
Availability details for
Quantity
  • Availability
  • Add to Cart