MAT238925040 Matrix Scientific

2-Amino-1-benzyl-4,5-dimethyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD01935932
  • Cas Number : 55817-72-6
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-155
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/042688.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042688.html
Availability details for
Quantity
  • Availability
  • Add to Cart