MAT238925041 Matrix Scientific

5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD01935935
  • Cas Number : 180903-14-4
  • Molecular Weight : 274.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-156
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042689.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042689.html
Availability details for
Quantity
  • Availability
  • Add to Cart