MAT238925060 Matrix Scientific

4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide

  • MDL number MFCD01568715
  • Cas Number : 341967-76-8
  • Molecular Weight : 283.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-181
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042708.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042708.html
Availability details for
Quantity
  • Availability
  • Add to Cart