MAT238925080 Matrix Scientific

1-(6-Methyl-2-pyridinyl)-1H-imidazole-4-carboxylic acid

  • MDL number MFCD02187274
  • Cas Number : 477847-81-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213-215
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042728.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042728.html
Availability details for
Quantity
  • Availability
  • Add to Cart