MAT238925087 Matrix Scientific

Potassium 4-iodobenzenesulfonate

  • MDL number MFCD06656232
  • Cas Number : 13035-63-7
  • Molecular Weight : 322.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042735.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042735.html
Availability details for
Quantity
  • Availability
  • Add to Cart