MAT238925092 Matrix Scientific

4-Acetyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD02571849
  • Cas Number : 16168-93-7
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-232
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/042740.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042740.html
Availability details for
Quantity
  • Availability
  • Add to Cart