MAT238925099 Matrix Scientific

3-(3-Methoxyphenyl)-1-oxo-3,4-dihydro-1H-isochromene-4-carboxylic acid

  • MDL number MFCD03001219
  • Cas Number : 400751-55-5
  • Molecular Weight : 298.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042747.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042747.html
Availability details for
Quantity
  • Availability
  • Add to Cart