MAT238925113 Matrix Scientific

Methyl 3-(4-bromophenyl)acrylate

  • MDL number MFCD00157162
  • Cas Number : 71205-17-9
  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042761.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042761.html
Availability details for
Quantity
  • Availability
  • Add to Cart