MAT238925176 Matrix Scientific

4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD06200910
  • Cas Number : 186956-98-9
  • Molecular Weight : 326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042824.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042824.html
Availability details for
Quantity
  • Availability
  • Add to Cart