MAT238925193 Matrix Scientific

Methyl 2-amino-5-(4-chlorophenyl)-1,3-thiazole-4-carboxylate

  • MDL number MFCD06200914
  • Cas Number : 127918-92-7
  • Molecular Weight : 268.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-240
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042841.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042841.html
Availability details for
Quantity
  • Availability
  • Add to Cart