MAT238925195 Matrix Scientific

Ethyl 2-(5-methoxy-1H-indol-3-yl)-2-oxoacetate

  • MDL number MFCD06200915
  • Cas Number : 14771-33-6
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042843.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042843.html
Availability details for
Quantity
  • Availability
  • Add to Cart