MAT238925229 Matrix Scientific

Methyl 5-iodo-6-oxo-1,6-dihydro-3-pyridinecarboxylate

  • MDL number MFCD08276965
  • Cas Number : 885950-46-9
  • Molecular Weight : 279.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-211
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042877.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042877.html
Availability details for
Quantity
  • Availability
  • Add to Cart