MAT238925276 Matrix Scientific

2-Methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid

  • MDL number MFCD00202110
  • Cas Number : 303148-48-3
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-150
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042925.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042925.html
Availability details for
Quantity
  • Availability
  • Add to Cart