MAT238925291 Matrix Scientific

3-(Benzylsulfanyl)-1H-1,2,4-triazol-5-ylamine

  • MDL number MFCD00141524
  • Cas Number : 3922-47-2
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-98
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042940.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042940.html
Availability details for
Quantity
  • Availability
  • Add to Cart