MAT238925299 Matrix Scientific

3-(2-Propynylsulfanyl)-1H-1,2,4-triazol-5-ylamine

  • MDL number MFCD00141528
  • Cas Number : 53918-44-8
  • Molecular Weight : 154.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042948.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042948.html
Availability details for
Quantity
  • Availability
  • Add to Cart