MAT238925305 Matrix Scientific

4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile

  • MDL number MFCD00794103
  • Cas Number : 477853-95-5
  • Molecular Weight : 291.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 330
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042954.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042954.html
Availability details for
Quantity
  • Availability
  • Add to Cart