MAT238925327 Matrix Scientific

3-Amino-4-[(2,6-dichlorophenyl)sulfanyl]-benzenecarbonitrile

  • MDL number MFCD01935934
  • Cas Number : 338982-10-8
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-184
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042976.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/042976.html
Availability details for
Quantity
  • Availability
  • Add to Cart