MAT238925337 Matrix Scientific

N-Hydroxy-4-methoxybenzenecarboximidoyl chloride

  • MDL number MFCD01924588
  • Cas Number : 38435-51-7
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/042986.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/042986.html
Availability details for
Quantity
  • Availability
  • Add to Cart