MAT238925354 Matrix Scientific

2-{[Imino(phenyl)methyl]amino}acetic acid

  • MDL number MFCD01935985
  • Cas Number : 32683-07-1
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043003.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043003.html
Availability details for
Quantity
  • Availability
  • Add to Cart