MAT238925356 Matrix Scientific

Ethyl 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD02083398
  • Cas Number : 111493-74-4
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043005.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043005.html
Availability details for
Quantity
  • Availability
  • Add to Cart