MAT238925358 Matrix Scientific

2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)-acetic acid

  • MDL number MFCD01935986
  • Cas Number : 338982-13-1
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-208
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043007.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043007.html
Availability details for
Quantity
  • Availability
  • Add to Cart