MAT238925395 Matrix Scientific

6-Phenyl-5-piperazino-3(2H)-pyridazinone

  • MDL number MFCD02571097
  • Cas Number : 132814-16-5
  • Molecular Weight : 256.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268-272
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043044.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043044.html
Availability details for
Quantity
  • Availability
  • Add to Cart