MAT238925408 Matrix Scientific

2-(3-Chloropropyl)-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

  • MDL number MFCD03001235
  • Cas Number : 385383-38-0
  • Molecular Weight : 373.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043057.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043057.html
Availability details for
Quantity
  • Availability
  • Add to Cart