MAT238925414 Matrix Scientific

1-(1,3-Thiazol-2-yl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD02665165
  • Cas Number : 383136-31-0
  • Molecular Weight : 178.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 135�m
Product Link http://www.matrixscientific.com/043063.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043063.html
Availability details for
Quantity
  • Availability
  • Add to Cart