MAT238925421 Matrix Scientific

Methyl 4-acetyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD02571735
  • Cas Number : 40611-82-3
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043070.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043070.html
Availability details for
Quantity
  • Availability
  • Add to Cart