MAT238925464 Matrix Scientific

1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04124379
  • Cas Number : 142818-02-8
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-117
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043113.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043113.html
Availability details for
Quantity
  • Availability
  • Add to Cart