MAT238925465 Matrix Scientific

Methyl 2-(1,2-benzisoxazol-3-yl)acetate

  • MDL number MFCD04117784
  • Cas Number : 59899-89-7
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/043114.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/043114.html
Availability details for
Quantity
  • Availability
  • Add to Cart